Absorbent socs, Allwik Universal
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
813789 813790
BRDY813789EA 110.6 EUR
BRDY813789 BRDY813790
Absorbent socs, Allwik Universal
Absorptiematerialen en bescherming tegen morsen
These Allwik-Socs are used for absorbing leaks and spills around machines.
  • Absorbs a variety of industrial liquids including aggressive chemicals
  • Flexible design allows the soc to fit around any machine, corner or curve
  • Filled with universal meltblown polypropylene