Absorbent pillows, Hazwik Chemical
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
813855 813853
BRDY813855EA 187.12 EUR
BRDY813855 BRDY813853
Absorbent pillows, Hazwik Chemical
Absorptiematerialen en bescherming tegen morsen
These Hazwik pillows are used for absorbing spills and leaking liquids in tight spaces.
  • Absorb chemicals, acids and bases
  • Good for large spills once contained
  • Ideal for use in packaging for extra protection during transport
  • Two different sizes to fit any application