Labels for BBP33 printers, type B-719
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
133875 134344 153178 153174 153181 153176 153182 153175 153177 153179 153180
BRDY133875EA 239.1 EUR
BRDY133875 BRDY134344 BRDY153178 BRDY153174 BRDY153181 BRDY153176 BRDY153182 BRDY153175 BRDY153177 BRDY153179 BRDY153180
Labels for BBP33 printers, type B-719
Etiketten
Static dissipative polyimide (B-719) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • Surface resistivity values in the recommended range for dissipative ESD packaging materials as defined by ANSI/ESD S541-2008
  • Service temperature of –70 to 100 °C
Low profile, high temperature, matte white polyimide material features a static dissipative adhesive and static dissipative release liner.
Certificaten: UL Approved; meets requirements of MIL-STD-202G, method 215K