Anti-slip floor tapes
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
078184 078189
BRDY078184EA 90.66 EUR
BRDY078184 BRDY078189
Anti-slip floor tapes
Tape
Anti-skid (anti-slip) tapes help to prevent slipping and falling on wet or slippery floors.
  • With stands −40 to 70 °C
  • Thickness of 0,66 mm
  • Permanent acrylic adhesive
The rugged laminated polyester tape is adhesive backed and coated with thousands of particles of durable aluminium oxide grit. Anti-Skid tape bonds permanently to clean, dry floors, withstands grease and oil and will work indoors or outdoors. Anti-Skid tape is available in a variety of colours and sizes to suit all of your floor marking needs.