Avantor® Hichrom HI-WAX HT, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI13-25-010-15EA 505 EUR
HI13-25-010-15 HI13-25-010-30 HI13-25-015-15 HI13-25-015-30 HI13-25-025-15 HI13-25-025-30 HI13-32-010-10 HI13-32-010-15 HI13-32-010-30 HI13-32-015-15 HI13-32-015-30 HI13-32-025-15 HI13-32-025-30 HI13-53-010-10 HI13-53-010-15 HI13-53-010-30 HI13-53-015-15 HI13-53-015-30 HI13-53-017-30 HI13-53-025-15 HI13-53-025-30 HI13-10-010-10 HI13-10-010-5 HIGCHI13-32-100-30
Avantor® Hichrom HI-WAX HT, GC Columns
Kolommen voor chromatografie
High Temperature unique PEG phase. Extend the temperature limits of your FAST-GC and GCxGC methods while using a polar WAX phase.
Max temperature limits may change depending on the film thickness.
Order Now
Specificaties
Phase name HI-WAX HT
Environmentally Preferable
Phase composition Polyethylene Glycol (PEG) - high temperature
Polarity Polar stationary phase
Crossbond Yes
Temperatuurlimiet 300 °C
USP code G14, G15, G16