Avantor® Hichrom HI-BASIC, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI38-25-015-25EA 1060 EUR
HI38-25-015-25 HI38-32-025-25 HI38-32-025-30 HIGCHI38-32-025-60
Avantor® Hichrom HI-BASIC, GC Columns
Kolommen voor chromatografie
Application specific column for basic compouns (e.g. amines).
Max temperature limits may change depending on the film thickness.