Avantor® Hichrom HI-DAI 1, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI42-25-250-30EA 1120 EUR
HI42-25-250-30 HI42-25-250-50 HI42-32-300-30 HI42-32-300-50 HI42-32-500-30 HI42-32-500-50
Avantor® Hichrom HI-DAI 1, GC Columns
Kolommen voor chromatografie
Application specifc column for direct introduction of aqueous samples, thus minimizing sample preparation.
Max temperature limits may change depending on the film thickness.