Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI71-25-015-15EA 873 EUR
HI71-25-015-15 HI71-25-015-25 HI71-25-015-30 HI71-25-015-50 HI71-25-015-60 HI71-25-025-15 HI71-25-025-25 HI71-25-025-30 HI71-25-025-50 HI71-25-025-60 HI83-10-010-10 HI83-10-010-5
Avantor® Hichrom HI-DEX DMT Beta, GC Columns
Kolommen voor chromatografie
Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.