Avantor® Hichrom HI-225 MS, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI23-18-018-10EA 528 EUR
HI23-18-018-10 HI23-18-018-20 HI23-18-018-40 HI23-18-018-50 HI23-18-018-60 HI23-20-010-10 HI23-20-010-20 HI23-20-010-40 HI23-20-010-50 HI23-20-010-60 HI23-20-020-10 HI23-20-020-20 HI23-20-020-40 HI23-20-020-50 HI23-20-020-60 HI23-25-010-15 HI23-25-010-30 HI23-25-010-50 HI23-25-010-60 HI23-25-025-10 HI23-25-025-105 HI23-25-025-15 HI23-25-025-30 HI23-25-025-50 HI23-25-025-60 HI23-32-010-10 HI23-32-010-105 HI23-32-010-15 HI23-32-010-30 HI23-32-010-50 HI23-32-010-60 HI23-32-025-10 HI23-32-025-105 HI23-32-025-15 HI23-32-025-30 HI23-32-025-50 HI23-32-025-60 HI23-53-010-10 HI23-53-010-15 HI23-53-010-30 HI23-53-025-15 HI23-53-025-30 HI23-53-025-50 HI23-53-025-60 HI23-05-005-2 HI23-05-005-2.5 HI23-05-005-5 HI23-10-010-10 HI23-10-010-15 HI23-10-010-5 HI23-15-015-10 HI23-15-015-15 HI23-15-015-20 HI23-15-015-30 HI23-15-015-5
Avantor® Hichrom HI-225 MS, GC Columns
Kolommen voor chromatografie
General purpose mid-to-high-polarity phase for a wide range of applications, like, for example: FAMEs, carbohydrates, sterols, flavor compounds. HI-225 MS is an MS grade crossbonded column.
  • Phase composition: Crossbonded 25% Cyanopropyl, 25% Phenyl, 50% Methyl Polysiloxane - MS
Max temperature limits may change depending on the film thickness.