Avantor® Hichrom HI-TCEP, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI61-25-040-30EA 748 EUR
HI61-25-040-30 HI61-25-040-60 HI61-25-044-60 HI61-32-051-60
Avantor® Hichrom HI-TCEP, GC Columns
Kolommen voor chromatografie
Application specifc columns ideal for aromatics and oxygenates in gasoline.
Max temperature limits may change depending on the film thickness.