Avantor® Hichrom HI-DEX G-03, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI75-25-015-15EA 1250 EUR
HI75-25-015-15 HI75-25-015-25 HI75-25-015-30 HI75-25-015-50 HI75-25-015-60 HI75-25-025-15 HI75-25-025-25 HI75-25-025-30 HI75-25-025-50 HI75-25-025-60 HI85-10-010-10 HI85-10-010-5
Avantor® Hichrom HI-DEX G-03, GC Columns
Kolommen voor chromatografie
Unique selectivity for the separation of enantiomers. Useful for the analysis of chiral compounds in fragrances, pesticides and pharmaceuticals in which the determination of the ratio of enantiomers in a sample is necessary.
Max temperature limits may change depending on the film thickness.