Avantor® Hichrom HI-17 HT, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI90-25-010-15EA 577 EUR
HI90-25-010-15 HI90-25-010-30 HI90-25-015-15 HI90-25-015-30 HI90-25-025-15 HI90-25-025-30 HI90-32-010-10 HI90-32-010-15 HI90-32-010-30 HI90-32-015-15 HI90-32-015-30 HI90-32-025-15 HI90-32-025-30 HI90-53-010-10 HI90-53-010-15 HI90-53-010-30 HI90-53-015-15 HI90-53-015-30 HI90-53-017-30 HI90-53-025-15 HI90-53-025-30 HI90-10-010-10 HI90-10-010-5
Avantor® Hichrom HI-17 HT, GC Columns
Kolommen voor chromatografie
High Temperature general purpose column. Ideal for confrmation analysis, ideal as high polarity dimension in GCxGC-HT confgurations.
Max temperature limits may change depending on the film thickness.