Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI41-32-100-30EA 1050 EUR
HI41-32-100-30 HI41-32-180-30
Avantor® Hichrom HI-BIODIESEL 110, GC Columns
Kolommen voor chromatografie
Application specifc column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Max temperature limits may change depending on the film thickness.