Avantor® Hichrom HI-SOLVE 2, GC Columns
Leverancier: Avantor
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI63-25-025-30EA 1090 EUR
HI63-25-025-30 HI63-32-025-50
Avantor® Hichrom HI-SOLVE 2, GC Columns
Kolommen voor chromatografie
Application specifc columns developed for complex solvents mixtures analysis. HI-SOLVE 2 is ideal for aromatics and oxygenates in gasoline.
Temperature limits may change depending on the film thickness.