Palette knife spatulas with handle, stainless steel
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A200-100 A200-200 A200-300 A201-100 A201-200 A201-300
300-0643EA 13.9 EUR
300-0643 300-0644 300-0645 300-0646 300-0647 300-0648
Palette knife spatulas with handle, stainless steel
Spatels
These pallet knives are ideal for work in laboratories and production. They are ideal for mixing, spreading or cutting material.
  • Stainless steel blade
  • Smooth handle
  • Robust design
  • Easy to use and clean
The handles are rounded so they are easy to use for long periods of time.