X-ray and metal detectable scrapers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SAMPM1019B-110EA 30.5 EUR
SAMPM1019B-110
X-ray and metal detectable scrapers
Schrapers
The X-ray and metal detectable scraper is ideal for scraping and mixing material. The scrapers are made from a special grade of PP that can be detected. The bright blue colour also means that they can be easily seen if left in the material in line with HACCP guidelines.
  • Robust PP construction, provides excellent chemical resistance to many products
  • Handy blade, simple to use
  • Full batch traceability
To ensure the cleanliness of the scrapers are manufactured and packed in a cleanroom. They can be used as a single use disposable product straight from the bag or reused.
Certificaten: These scrapers conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Verpakking: Supplied individually bagged.