X-ray and metal detectable jugs, PP, graduated
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1044B-1000 M1044B-500
SAMPM1044B-1000EA 69.7 EUR
SAMPM1044B-1000 SAMPM1044B-500
X-ray and metal detectable jugs, PP, graduated
Bekers
X-ray and metal detectable jugs are perfect for both laboratory and production work. They are available in 2 sizes. The bright blue colour makes the jugs easy to spot if they get left in the product.
  • Robust PP construction
  • Excellent chemical resistance
  • Non sterile
  • Full batch traceability
To ensure the cleanliness of the jugs, they are manufactured and packed in a cleanroom. The robust nature of these jugs means that they can give many years of service or for more demanding applications. They can be used as a single use disposable product straight from the bag.
Ideal for use in food, pharma, chemical industries and use with powders, granules and pastes.
Certificaten: These PP jugs conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
Verpakking: Supplied individually bagged.