Foetal bovine serum
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S1400-500 S1400-050 S1400-100 S140H-050 S140H-100 S140H-500 S140F-050 S140F-100 S140F-500
S1400-500EA 357 EUR
S1400-500 S1400-050 S1400-100 S140H-050 S140H-100 S140H-500 S140F-050 S140F-100 S140F-500
Foetal bovine serum
Serum
High quality, sterile filtered foetal bovine serum (FBS) derived exclusively from EU countries. Each manufactured batch is rigorously controlled, from the collection of serum through to final packaging.
  • EU origin
  • High quality serum, rigorously tested
  • Full traceability and security
  • Naturally lower IgG level
  • More ecological transport