Slide storing system
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
945L1 946L1 947L1 948L1
631-1283EA 1140 EUR
631-1283 631-1284 631-1285 631-1286
Slide storing system
Objectglaasjeshouders
Made from high impact PMMA and steel, this slide storage system is designed for secure classification and storage of microscope slides.
  • Lightweight and functional
  • Durable steel frame for stability
  • Semi-transparent, ice coloured drawers provide an attractive finish
  • Transparent lids permit easy drawer viewing, while protecting slides from dust and foreign bodies
  • System holds up to 1300 slides per drawer, 5200 slides in total
The system is composed of 4 sliding drawers, a closing cover, and 4 fixed support feet. As an alternative, the system is also available with 4 swivel castors for added mobility. Each single drawer can hold up to 13 slide supports and transparent cover plates (must be ordered separately). Slide supports are easily removed, and will accommodate up to 100 microscope slides. Each cavity is numbered. The complete storing system can hold up to 5200 slides.