Upright microscopes, DM 1000 LED
Leverancier: LEICA MICROSYSTEMS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
11888842_P1 11888842_P2 11888842_P3 11888842_P4 11888842_P5 11888842_P6 11888842_P7 11888842_P8
630-3168EA 5690 EUR
630-3168 630-3169 630-3170 630-3171 630-3172 630-3173 630-3174 630-3175
Upright microscopes, DM 1000 LED
Microscopen
The DM1000 LED laboratory microscope is perfect for clinical laboratory applications; histology, cytology, haematology and pathology.
  • Compatible with the full line of Leica microscope optics and accessories for increased flexibility
  • Choice of traditional Tungsten Halogen light source or LED illumination that makes changing lamps a thing of the past
  • Appropriately scaled footprint to meet the needs of even the most space challenged laboratory
  • Easily the best all round choice in microscopes for transmitted light applications; brightfield and phase contrast
LED features a long life LED that provides near daylight, bright illumination with constant colour temperature and emits less heat.
Designed for occasional use or the rigorous demands of routine laboratory use, incorporating ergonomics that are based on the natural movements of the user. Height adjustable focus knobs provide the optimal choice for a relaxed hand position. Adjustable eyetubes promote correct posture. Symmetrical operation from either left or right facilitates alternating users. Telescopic coaxial stage control with friction adjustment for user selected resistance. Specially designed Ergolift where both pitch and height can be adjusted to meet the needs of users.
Bestelinformatie: 2 year warranty.
Leveringsinformatie: Package I includes: Leica DM1000 LED, Koehler Kit for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser CL/PH (0.90/1.25 Oil, S1, CC), ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Standard tube HC -/4/4, two Eyepieces HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Objective HI PLAN 10x/0.25, Objective HI PLAN 20x/0.40, 0.17, 0.98, Objective HI PLAN 40x/0,65, Objective HI PLAN 100x/1,25 oil, Immersion oil (7,4 ml).
Package II includes: Leica DM1000 LED, Koehler Kit for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser CL/PH (0.90/1.25 Oil, S1, CC), ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Phototube HC L1T 4/5/7 (trinocular), two Eyepieces HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Objective HI PLAN 10x/0.25, Objective HI PLAN 20x/0.40 (0.17, 0.98), Objective HI PLAN 40x/0,65, Objective HI PLAN 100x/1,25 oil, Immersion oil (7,4 ml).
Package III includes: Leica DM1000 LED, Koehler Kit for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser UCL 0.90/1.25 OIL S1, Set of light rings UCL DF (PH 1/2/3), ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Standard tube HC -/4/4, two Eyepieces HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Obj. HI PLAN 10x/0.25 PH1, Obj. HI PLAN 20x/0.40 PH1, Obj. HI PLAN 40x/0.65 PH2, Obj. HI PLAN 100x/1.25 OIL PH3, Immersion oil (7,4 ml).
Package IV includes: Leica DM1000 LED, Koehler Kit for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser UCL 0.90/1.25 OIL S1, Set of light rings UCL DF (PH 1/2/3), ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Phototube HC L1T 4/5/7 (trinocular), two Eyepieces HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Obj. HI PLAN 10x/0.25 PH1, Obj. HI PLAN 20x/0.40 PH1, Obj. HI PLAN 40x/0.65 PH2, Obj. HI PLAN 100x/1.25 OIL PH3, Immersion oil (7,4 ml).
Package V includes: Leica DM1000 LED, Fix Koehler for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser CL/PH 0.90/1.25 (Oil, S1, CC), ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Standard tube HC -/4/4, Eyepiece HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Objective HI PLAN 10x/0.25, Objective HI PLAN 20x/0.40, 0.17, 0.98, Objective HI PLAN 40x/0,65, Objective HI PLAN 100x/1,25 oil, Immersion oil (7,4 ml).
Package VI includes: Leica DM1000 LED, Fix Koehler for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser CL/PH 0.90/1.25 (Oil, S1, CC), ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Phototube HC L1T 4/5/7 (trinocular), Eyepiece HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Objective HI PLAN 10x/0.25, Objective HI PLAN 20x/0.40, 0.17, 0.98, Objective HI PLAN 40x/0,65, Objective HI PLAN 100x/1,25 oil, Immersion oil (7,4 ml).
Package VII includes: Leica DM1000 LED, Fix Koehler for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser UCL 0.90/1.25 OIL S1, Set of light rings UCL DF,PH 1/2/3, ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Standard tube HC -/4/4, Eyepiece HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Obj. HI PLAN 10x/0.25 PH1, Obj. HI PLAN 20x/0.40 PH1, Obj. HI PLAN 40x/0.65 PH2, Obj. HI PLAN 100x/1.25 OIL PH3, Immersion oil (7,4 ml).
Package VIII includes: Leica DM1000 LED, Fix Koehler for DM1000, Focus knobs for DM1000, Dust cover for DM1000 (DM LS/LSP), Condenser lens CL/LS, Condenser UCL 0.90/1.25 OIL S1, Set of light rings UCL DF,PH 1/2/3, ErgoStage with Ceramic Plate for DM1000, Slide holder one hand slide change, Object clamp for oil immersion, XY-stage control standard, Phototube HC L1T 4/5/7 (trinocular), Eyepiece HC PLAN s 10x/22 Br. M, Objective HI PLAN 4x/0.10, Obj. HI PLAN 10x/0.25 PH1, Obj. HI PLAN 20x/0.40 PH1, Obj. HI PLAN 40x/0.65 PH2, Obj. HI PLAN 100x/1.25 OIL PH3, Immersion oil (7,4 ml).