Foetal bovine serum, EU origin, Premium
Leverancier: Biowest
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
S140B-100 S140B-500 S140B-050
S140B-100EA 85.5 EUR
S140B-100 S140B-500 S140B-050
Foetal bovine serum, EU origin, Premium
Serum
In ordering FBS Premium, you can avoid time consuming batch testing and batch reservations.
  • Sterile filtered
  • Frozen
  • Storage at –20 °C
  • Shelf life of 60 months
  • Shipping conditions: –20 °C or dry ice
FBS Premium is a selection of the best batches, selected on excellent values of Endotoxin, Hemoglobin, and Cell growth on 4 Cell lines: SP2/O-Ag14, HELA, L929 and MRC-5, guaranteeing your success in cell culture.