Tube clamps, Pure-Fit® TC
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PFTC375 PFTC750 PFTC750PVDF PFTC375PVDF
VERNPFTC375EA 224 EUR
VERNPFTC375 VERNPFTC750 VERNPFTC750PVDF VERNPFTC375PVDF
Tube clamps, Pure-Fit® TC
Klemmen Slangklemmen
Pure-Fit® TC tube clamps represent a new generation of clamping technology for the pharmaceutical industry.
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Custom colours and identification available
  • Autoclavable and sterilisable (autoclaving not recommended for PP clamps)
They feature an outer body with no sharp edges, eliminating the potential for snags, scratches and punctures; a unique press-down locking system that is fully encapsulated to guard against untimely or unwanted release; a side release mechanism that facilitates secure, single-handed installation and operation; and an integral bore hole for absolute lock-out capabilities. Pure-Fit TC’s innovative design allows for complete flow stoppage, and the clamp can be installed over the tube in the normal fashion or put together after the tube is already part of a completed assembly.
Pure-Fit® TC tube clamps are constructed from animal derivative-free high performance thermoplastic materials (PVDF or PP), and come in two standard sizes (⅜" and ¾") to accommodate a wide range of tubing products.
Certificaten: Meets USP Class VI criteria.