Tube clamps, Pure-Fit® TCL
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PFTCL150 PFTCL250
VERNPFTCL150EA 2220 EUR
VERNPFTCL150 VERNPFTCL250
Tube clamps, Pure-Fit® TCL
Klemmen Slangklemmen
Pure-Fit® TCL large tube clamps provide the same innovative clamping technology as Pure-Fit TC but are designed to accommodate large diameter and thick wall tubing applications.
  • Smooth edges and corners help prevent punctures and ruptures
  • Can be installed on or over tubing or finished assemblies
  • High degree of leverage accommodates tubing with different durometers
  • Custom colours and identification available
  • Fully autoclavable and sterilisable
Pure-Fit TCL is compatible with standard, braided (reinforced) and multilayer tubing. In addition to providing complete fluid stoppage, Pure-Fit TCL can be used in the flow position and features an integral bore hole for absolute lock-out capabilities.
Pure-Fit® TCL is constructed from animal derivative-free PVDF. Pure-Fit TCL is available in two sizes to accommodate a wide range of thick wall tubing and can be used with up to 2,0" O.D. tubing.
Certificaten: Meets USP Class VI criteria.