Bismuth ICP Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1010-100ML E1137-100ML
HONCE1010-100MLEA 104 EUR
HONCE1010-100ML HONCE1137-100ML
Bismuth ICP Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Bismut standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).