Cesium ICP standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1016-100ML E1105-100ML E1143-100ML E1213-100ML
HONCE1016-100MLEA 79.2 EUR
HONCE1016-100ML HONCE1105-100ML HONCE1143-100ML HONCE1213-100ML
Cesium ICP standards, (CRMs), Fluka™
Standaarden Kationische standaarden Cesium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).