Holmium ICP standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1027-100ML E1154-100ML
HONCE1027-100MLEA 157 EUR
HONCE1027-100ML HONCE1154-100ML
Holmium ICP standards, (CRMs), Fluka™
Standaarden Kationische standaarden Holmium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).