Thallium ICP Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1064-100ML
E1191-100ML
HONCE1064-100MLEA
79.2
EUR
HONCE1064-100ML
HONCE1191-100ML
Thallium ICP Standards, (CRMs), Fluka™
Standaarden
Kationische standaarden
Thallium standaarden
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).