Yttrium ICP Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1069-100ML E1196-100ML
HONCE1069-100MLEA 79.2 EUR
HONCE1069-100ML HONCE1196-100ML
Yttrium ICP Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Yttrium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).