Zinc ICP standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1071-100ML E1128-100ML E1198-100ML E1265-100ML
HONCE1071-100MLEA 79.2 EUR
HONCE1071-100ML HONCE1128-100ML HONCE1198-100ML HONCE1265-100ML
Zinc ICP standards, (CRMs), Fluka™
Standaarden Kationische standaarden Zink standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).