Palladium ICP and ICP-MS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1340-100ML E1408-100ML
HONCE1340-100MLEA 202 EUR
HONCE1340-100ML HONCE1408-100ML
Palladium ICP and ICP-MS standards, (CRMs), Fluka™
Standaarden Kationische standaarden Palladium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Order Now
Specificaties
Environmentally Preferable