Tantalum ICP & ICP-MS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1355-100ML
E1423-100ML
HONCE1355-100MLEA
113
EUR
HONCE1355-100ML
HONCE1423-100ML
Tantalum ICP & ICP-MS standards, (CRMs), Fluka™
Standaarden
Kationische standaarden
Tantalum standaarden
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).