Uranium ICP and ICP-MS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1361-100ML
E1430-100ML
HONCE1361-100MLEA
139
EUR
HONCE1361-100ML
HONCE1430-100ML
Uranium ICP and ICP-MS Standards, (CRMs), Fluka™
Standaarden
Kationische standaarden
Uranium standaarden
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).