Tungsten ICP and ICP-MS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1363-100ML E1432-100ML
HONCE1363-100MLEA 113 EUR
HONCE1363-100ML HONCE1432-100ML
Tungsten ICP and ICP-MS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Wolfraam standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).