Arsenic(V) ICP standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E1501-100ML
E1505-100ML
HONCE1501-100MLEA
123
EUR
HONCE1501-100ML
HONCE1505-100ML
Arsenic(V) ICP standards, (CRMs), Fluka™
Standaarden
Kationische standaarden
Arseen standaarden
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).