Chromium AAS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4019-100ML E4019-500ML E4020-100ML E4020-500ML
HONCE4019-100MLEA 34.7 EUR
HONCE4019-100ML HONCE4019-500ML HONCE4020-100ML HONCE4020-500ML
Chromium AAS standards, (CRMs), Fluka™
Standaarden Kationische standaarden Chroom standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).