Manganese AAS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4044-100ML E4044-500ML E4045-100ML E4045-500ML
HONCE4044-100MLEA 34.7 EUR
HONCE4044-100ML HONCE4044-500ML HONCE4045-100ML HONCE4045-500ML
Manganese AAS standards, (CRMs), Fluka™
Standaarden Kationische standaarden Mangaan standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).