Niobium AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4051-100ML E4051-500ML
HONCE4051-100MLEA 0 EUR
HONCE4051-100ML HONCE4051-500ML
Niobium AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Niobium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).