Neodymium AAS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4052-100ML E4052-500ML
HONCE4052-100MLEA 47.7 EUR
HONCE4052-100ML HONCE4052-500ML
Neodymium AAS standards, (CRMs), Fluka™
Standaarden Kationische standaarden Neodymium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).