Lead AAS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4055-100ML E4055-500ML E4115-100ML
HONCE4055-100MLEA 34.3 EUR
HONCE4055-100ML HONCE4055-500ML HONCE4115-100ML
Lead AAS standards, (CRMs), Fluka™
Standaarden Kationische standaarden Lood standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).