Rhenium AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4060-100ML E4060-500ML
HONCE4060-100MLEA 252 EUR
HONCE4060-100ML HONCE4060-500ML
Rhenium AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Renium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).