Rhodium AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4061-100ML
E4061-500ML
HONCE4061-100MLEA
408
EUR
HONCE4061-100ML
HONCE4061-500ML
Rhodium AAS Standards, (CRMs), Fluka™
Standaarden
Kationische standaarden
Rodium standaarden
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).