Ruthenium AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4062-100ML E4062-500ML
HONCE4062-100MLEA 164 EUR
HONCE4062-100ML HONCE4062-500ML
Ruthenium AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Ruthenium standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).