Tin AAS Standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4070-100ML E4070-500ML E4071-100ML E4071-500ML
HONCE4070-100MLEA 34.9 EUR
HONCE4070-100ML HONCE4070-500ML HONCE4071-100ML HONCE4071-500ML
Tin AAS Standards, (CRMs), Fluka™
Standaarden Kationische standaarden Tin standaarden
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).