Cerium AAS standards, (CRMs), Fluka™
Leverancier: Honeywell Chemicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E4016-100ML
E4016-500ML
HONCE4016-100MLEA
42.1
EUR
HONCE4016-100ML
HONCE4016-500ML
Cerium AAS standards, (CRMs), Fluka™
Standaarden
Kationische standaarden
Cerium standaarden
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certificaten: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).