VWR® ONEBAG®, Dehydrated buffered peptone water in bags with reconstitution water
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
710-2049EA 96.1 EUR
710-2049 710-2050
VWR® ONEBAG®, Dehydrated buffered peptone water in bags with reconstitution water
Media voor microbiologie Geprepareerde media voor microbiologie
Ready to use buffered peptone water is prepared a few minutes before use, to guarantee the best performance of the medium.
  • Long shelf life: 24 months at room temperature
  • Bag, water and culture medium sterilised by gamma irradiation (no over-heating; better performance of the medium)
  • No need for dispensing equipment (eg. diluter, deionised water producer and more)
  • Culture media is prepared in a few minutes before use, to guaranteee the best performance of the medium
Available to make up 90 or 225 ml of medium.
Certificaten: In compliance with ISO 6887, ISO 6579 and ISO 11290-2.