VWR® Gentle ReView™, Stripping Buffer
Leverancier: VWR ChemicalsTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Gentle ReView™ Stripping Buffer is a mild, yet effective method for stripping primary and secondary antibodies from PVDF or nitrocellulose membranes.
- Odor-free, room temperature procedure
- Minimize loss of antigen
The gentle nature of this buffer makes it possible to re-probe the same membrane several times without damaging the membrane-bound antigen. It is an odorless, ready-to-use formula that equires no mixing or heating prior to use. Most antibody-antigen complexes can be dissociated within 30 minutes at room temperature by Gentle ReView™ Stripping Buffer. However, incubation times and temperatures are dependent upon the affinity of the antibody-antigen interaction and may need to be optimized for each combination used.
Opgelet: For research use only. Not for therapeutic or diagnostic use.