Test tube racks, 50 ml
Leverancier: WDF - Funke GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2014-26-11 2014-26-13 2014-24-13 2014-25-11 2014-25-13
216-7240EA 0 EUR
216-7240 WCDF2014-26-13 WCDF2014-24-13 WCDF2014-25-11 WCDF2014-25-13
Test tube racks, 50 ml
Rekken Tube Racks
Racks for 50 ml test tubes.
5 mm space between holes.