Diffusor for ring light, VisiLED, S40-75
Leverancier: Schott AG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
VisiLED
SCOC400580EA 327 EUR
SCOC400580
Diffusor for ring light, VisiLED, S40-75
Lampen Ringlichten
The diffusor for LED ring lights avoids reflections of LEDs for uniform illumination.
  • Diffusor with adapting thread (M110×1) to avoid reflections of LEDs
Designed for use only with VisiLED ringlight S40-75.
Bestelinformatie: VisiLED ringlight S40-75 and controller MC 750, MC 1100, or MC 1500 required.