Waterstofperoxide 30 - 32%%, NORMATOM® for trace metal analysis
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
85961.260EA 128.4 EUR
85961.260 85961.290
Waterstofperoxide 30 - 32%%, NORMATOM® for trace metal analysis
Waterstofperoxide
Formule: H₂O₂
MW: 34.01 g/mol
Kookpunt: ~107 °C (1013 hPa)
Smeltpunt: -0.41
Dichtheid: 1.11 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011333
CAS nummer: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8) II
Merck Index: 14,04798
Order Now
Specification Test Results
Assay (H₂O₂) 30 - 32% w/w
Colouration max. 10 APHA
Cl (Chloride) max. 0.2 ppb
Ag (Silver) max. 0.1 ppb
Al (Aluminum) max. 1 ppb
As (Arsenic) max. 0.5 ppb
Au (Gold) max. 0.1 ppb
Ba (Barium) max. 0.1 ppb
Be (Beryllium) max. 0.1 ppb
Bi (Bismuth) max. 0.1 ppb
Ca (Calcium) max. 1 ppb
Cd (Cadmium) max. 0.5 ppb
Ce (Cerium) max. 0.1 ppb
Co (Cobalt) max. 0.5 ppb
Cr (Chromium) max. 1 ppb
Cs (Cesium) max. 0.1 ppb
Cu (Copper) max. 0.5 ppb
Dy (Dysprosium) max. 0.1 ppb
Er (Erbium) max. 0.1 ppb
Eu (Europium) max. 0.1 ppb
Fe (Iron) max. 1 ppb
Ga (Gallium) max. 0.1 ppb
Gd (Gadolinium) max. 0.1 ppb
Ge (Germanium) max. 0.1 ppb
Hf (Hafnium) max. 0.1 ppb
Hg (Mercury) max. 0.1 ppb
Ho (Holmium) max. 0.1 ppb
In (Indium) max. 0.1 ppb
K (Potassium) max. 1 ppb
La (Lanthanum) max. 0.1 ppb
Li (Lithium) max. 0.1 ppb
Lu (Lutetium) max. 0.1 ppb
Mg (Magnesium) max. 1 ppb
Mn (Manganese) max. 0.1 ppb
Mo (Molybdenum) max. 0.1 ppb
Na (Sodium) max. 1 ppb
Nb (Niobium) max. 0.1 ppb
Nd (Neodymium) max. 0.1 ppb
Ni (Nickel) max. 0.5 ppb
Pb (Lead) max. 0.1 ppb
Pd (Palladium) max. 0.5 ppb
Pr (Praseodymium) max. 0.1 ppb
Pt (Platinum) max. 0.5 ppb
Rb (Rubidium) max. 0.1 ppb
Re (Rhenium) max. 0.1 ppb
Rh (Rhodium) max. 0.5 ppb
Ru (Ruthenium) max. 0.5 ppb
Sb (Antimony) max. 0.5 ppb
Sc (Scandium) max. 0.1 ppb
Se (Selenium) max. 1 ppb
Sm (Samarium) max. 0.1 ppb
Sn (Tin) max. 0.5 ppb
Sr (Strontium) max. 0.1 ppb
Ta (Tantalum) max. 0.1 ppb
Tb (Terbium) max. 0.1 ppb
Te (Tellurium) max. 0.1 ppb
Th (Thorium) max. 0.1 ppb
Ti (Titanium) max. 0.5 ppb
Tl (Thallium) max. 0.1 ppb
Tm (Thulium) max. 0.1 ppb
U (Uranium) max. 0.1 ppb
V (Vanadium) max. 0.5 ppb
W (Tungsten) max. 0.1 ppb
Y (Yttrium) max. 0.1 ppb
Yb (Ytterbium) max. 0.1 ppb
Zn (Zinc) max. 0.5 ppb
Zr (Zirconium) max. 0.1 ppb