Funnels, with sintered glass filter plate
Leverancier: Kavalier
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
632424334103 632424334204 632424334206 632424334303 632424334304 632424334306 632424334412
KAVA632424334103EA 1110 EUR
KAVA632424334103 KAVA632424334204 KAVA632424334206 511-0668 KAVA632424334304 KAVA632424334306 511-0669
Funnels, with sintered glass filter plate
Trechters Filtertrechters Filtertrechters
Made from borosilicate 3.3 glass. With integrated sintered glass filter plates of various porosities.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, high resistance to temperature changes
Limited Time Sale
Select items below are currently on sale